Tisk…

5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride

OR18276-5G OR18276-25G OR18276-100G
APOSOR18276-5GEA 622 CZK
APOSOR18276-5G APOSOR18276-25G APOSOR18276-100G
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Vzorec: C₄H₇ClN₄O
MW: 162,58 g/mol
Bod tání: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00012704
CAS číslo: 72-40-2
EINECS: 200-778-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR