2-Chloro-1-iodo-5-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Chloro-2-iodo-4-nitrobenzene, 4-Chloro-3-iodonitrobenzene
OR18059-1G
OR18059-5G
OR18059-25G
APOSOR18059-1GEA
622
CZK
APOSOR18059-1G
APOSOR18059-5G
APOSOR18059-25G
2-Chloro-1-iodo-5-nitrobenzene
2-Chloro-1-iodo-5-nitrobenzene
Vzorec:
C₆H₃ClINO₂ MW: 283.4500 g/mol Bod tání: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD11521301 CAS číslo: 74534-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...