2,5-Dibromothieno[3,2-b]thiophene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR18024-5G
APOSOR18024-5GEA
4140
CZK
APOSOR18024-5G
2,5-Dibromothieno[3,2-b]thiophene 98%
2,5-Dibromothieno[3,2-b]thiophene
Vzorec:
C₆H₂Br₂S₂ MW: 298,0181 g/mol Bod tání: 124…129 °C Storage Temperature: Freezer |
MDL Number:
MFCD03931293 CAS číslo: 25121-87-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...