2-Benzylacrylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methylene-3-phenylpropionic Acid
OR18022-100G
OR18022-25G
APOSOR18022-100GEA
1240
CZK
APOSOR18022-100G
APOSOR18022-25G
2-Benzylacrylic acid
2-Benzylacrylic acid
Vzorec:
C₁₀H₁₀O₂ MW: 162,19 g/mol Bod tání: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00854174 CAS číslo: 5669-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...