2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Hydroxy-2-(methylthio)pyrimidin-4(3H)-one, 4,6-Dihydroxy-2-methylsulphanylpyrimidine, 4-Hydroxy-2-(methylthio)pyrimidin-6(1H)-one
OR17939-500G
APOSOR17939-500GEA
3110
CZK
APOSOR17939-500G
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98+%
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione
Vzorec:
C₅H₆N₂O₂S MW: 158,18 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00810414 CAS číslo: 1979-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...