Tisk…

3-Amino-2-bromobenzoic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR17935-1G OR17935-25G OR17935-5G
APOSOR17935-1GEA 622 CZK
APOSOR17935-1G APOSOR17935-25G APOSOR17935-5G
3-Amino-2-bromobenzoic acid
3-Amino-2-bromobenzoic acid
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
Bod tání: 153…158 °C
Storage Temperature: Ambient
MDL Number: MFCD07786957
CAS číslo: 168899-61-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR