Tisk…

1-Aminopyridinium iodide 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Aminopyridinium Iodide, 1-Aminopyridine hydroiodide, 1-Aminopyridine HI

OR17820-100G OR17820-25G
APOSOR17820-100GEA 6190 CZK
APOSOR17820-100G APOSOR17820-25G
1-Aminopyridinium iodide 98%
1-Aminopyridinium iodide
Vzorec: C₅H₇IN₂
MW: 222,03 g/mol
Bod tání: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051893
CAS číslo: 6295-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR