Tisk…

Phenothiazine 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 10H-Phenothiazine

OR17816-250G OR17816-500G OR17816-5KG
APOSOR17816-250GEA 2480 CZK
APOSOR17816-250G APOSOR17816-500G APOSOR17816-5KG
Phenothiazine 99%
Phenothiazine
Vzorec: C₁₂H₉NS
MW: 199,28 g/mol
Bod varu: 371 °C
Bod tání: 182…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005015
CAS číslo: 92-84-2
EINECS: 202-196-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR