2-Naphthyl boronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
OR1779-25G
OR1779-5G
OR1779-100G
APOSOR1779-25GEA
910
CZK
APOSOR1779-25G
APOSOR1779-5G
APOSOR1779-100G
2-Naphthyl boronic acid
2-Naphthyl boronic acid
Vzorec:
C₁₀H₉BO₂ MW: 171,99 g/mol Bod tání: 267…268 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236051 CAS číslo: 32316-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...