Tisk…

1-Naphthyl boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

OR1778-100G OR1778-25G
APOSOR1778-100GEA 2480 CZK
APOSOR1778-100G APOSOR1778-25G
1-Naphthyl boronic acid
1-Naphthyl boronic acid
Vzorec: C₁₀H₉BO₂
MW: 171,99 g/mol
Bod tání: 204…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00019722
CAS číslo: 13922-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR