Tisk…

Quinoxaline-6-acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Quinoxalin-6-yl)acetic acid

OR17707-1G OR17707-250MG
APOSOR17707-1GEA 10820 CZK
APOSOR17707-1G APOSOR17707-250MG
Quinoxaline-6-acetic acid
Quinoxaline-6-acetic acid
Vzorec: C₁₀H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09880609
CAS číslo: 473932-16-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR