4-Methyl-1-piperazineacetic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR17668-10G
OR17668-1G
APOSOR17668-10GEA
4200
CZK
APOSOR17668-10G
APOSOR17668-1G
4-Methyl-1-piperazineacetic acid 97%
4-Methyl-1-piperazineacetic acid
Vzorec:
C₇H₁₄N₂O₂ MW: 158,1983 g/mol Bod tání: 130…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD03444554 CAS číslo: 54699-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...