Tisk…

4-Bromo-6-methylisoquinoline

Dodavatel: Apollo Scientific
Varování

OR17654-1G OR17654-250MG
APOSOR17654-1GEA 2530 CZK
APOSOR17654-1G APOSOR17654-250MG
4-Bromo-6-methylisoquinoline
4-Bromo-6-methylisoquinoline
Vzorec: C₁₀H₈BrN
MW: 222,0812 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922845
CAS číslo: 1204298-52-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR