3-Bromo-2-methoxyaniline
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-o-anisidine, 2-Amino-6-bromoanisole
OR17601-1G
OR17601-5G
OR17601-10G
APOSOR17601-1GEA
622
CZK
APOSOR17601-1G
APOSOR17601-5G
APOSOR17601-10G
3-Bromo-2-methoxyaniline
3-Bromo-2-methoxyaniline
Vzorec:
C₇H₈BrNO MW: 202,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13186737 CAS číslo: 116557-46-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...