Tisk…

7-Quinolinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

OR17564-1G OR17564-250MG OR17564-5G
APOSOR17564-1GEA 2730 CZK
APOSOR17564-1G APOSOR17564-250MG APOSOR17564-5G
7-Quinolinecarboxylic acid
7-Quinolinecarboxylic acid
Vzorec: C₁₀H₇NO₂
MW: 173.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00047617
CAS číslo: 1078-30-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR