7-Quinolineboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Quinoline-7-boronic acid, 7-Quinolinylboronic acid
OR17562-1G
OR17562-250MG
APOSOR17562-1GEA
5920
CZK
APOSOR17562-1G
APOSOR17562-250MG
7-Quinolineboronic acid 95%
7-Quinolineboronic acid
Vzorec:
C₉H₈BNO₂ MW: 172.9800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11100750 CAS číslo: 629644-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...