Tisk…

7-Bromoquinoline

Dodavatel: Apollo Scientific
Varování

OR17561-25G OR17561-5G OR17561-1G
APOSOR17561-25GEA 2480 CZK
APOSOR17561-25G APOSOR17561-5G APOSOR17561-1G
7-Bromoquinoline
7-Bromoquinoline
Vzorec: C₉H₆BrN
MW: 208,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03695823
CAS číslo: 4965-36-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR