Tisk…

2,5-Dibromo-4-methylthiazole

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,5-Dibromo-4-methyl-1,3-thiazole

OR17553-1G OR17553-250MG
APOSOR17553-1GEA 992 CZK
APOSOR17553-1G APOSOR17553-250MG
2,5-Dibromo-4-methylthiazole
2,5-Dibromo-4-methylthiazole
Vzorec: C₄H₃Br₂NS
Storage Temperature: Refrigerator
MDL Number: MFCD09260910
CAS číslo: 79247-78-2
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR