Tisk…

5-Bromo-4-methylisoquinoline

Dodavatel: Apollo Scientific
Varování

OR17552-1G OR17552-500MG
APOSOR17552-1GEA 4440 CZK
APOSOR17552-1G APOSOR17552-500MG
5-Bromo-4-methylisoquinoline
5-Bromo-4-methylisoquinoline
Vzorec: C₁₀H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD12922846
CAS číslo: 651310-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR