2-Bromo-5-chloro-3-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-5-chloro-3-picoline
OR17542-1G
OR17542-5G
APOSOR17542-1GEA
622
CZK
APOSOR17542-1G
APOSOR17542-5G
2-Bromo-5-chloro-3-methylpyridine
2-Bromo-5-chloro-3-methylpyridine
Vzorec:
C₆H₅BrClN MW: 206.47 Bod tání: 36…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095062 CAS číslo: 65550-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...