6-Chloro-4-cyclopropyl-1H-quinazolin-2-one 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-4-cyclopropylquinazolin-2(1H)-one
OR17533-100MG
OR17533-250MG
APOSOR17533-100MGEA
1450
CZK
APOSOR17533-100MG
APOSOR17533-250MG
6-Chloro-4-cyclopropyl-1H-quinazolin-2-one 95+%
6-Chloro-4-cyclopropyl-1H-quinazolin-2-one
Vzorec:
C₁₁H₉ClN₂O MW: 220,66 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11109434 CAS číslo: 150878-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...