Tisk…

(1-Acetylpiperidin-4-yl)acetic acid

Dodavatel: Apollo Scientific

OR17507-1G OR17507-5G
APOSOR17507-1GEA 5630 CZK
APOSOR17507-1G APOSOR17507-5G
(1-Acetylpiperidin-4-yl)acetic acid
(1-Acetylpiperidin-4-yl)acetic acid
Vzorec: C₉H₁₅NO₃
MW: 185,22 g/mol
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD12027293
CAS číslo: 885274-84-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR