Tisk…

a-Tosylbenzyl isocyanide 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (1-Phenyl-1-tosyl)methyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide

OR1744-1G OR1744-5G
APOSOR1744-1GEA 2240 CZK
APOSOR1744-1G APOSOR1744-5G
a-Tosylbenzyl isocyanide 97%
a-Tosylbenzyl isocyanide
Vzorec: C₁₅H₁₃NO₂S
MW: 271,3342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114776
CAS číslo: 36635-66-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR