Tisk…

4-Methoxyanthranilic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-4-methoxybenzoic acid

OR17403-25G OR17403-1G OR17403-5G
APOSOR17403-25GEA 1870 CZK
APOSOR17403-25G APOSOR17403-1G APOSOR17403-5G
4-Methoxyanthranilic acid
4-Methoxyanthranilic acid
Vzorec: C₈H₉NO₃
MW: 167,16 g/mol
Bod tání: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00667729
CAS číslo: 4294-95-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR