2-Aminopyridine-4-boronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Aminopyridine-4-boronic acid, pinacol ester, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR17204-1G
OR17204-100MG
OR17204-5G
APOSOR17204-1GEA
6850
CZK
APOSOR17204-1G
APOSOR17204-100MG
APOSOR17204-5G
2-Aminopyridine-4-boronic acid pinacol ester
2-Aminopyridine-4-boronic acid pinacol ester
Vzorec:
C₁₁H₁₇BN₂O₂ MW: 220,08 g/mol Bod tání: 161…162 °C Storage Temperature: Freezer |
MDL Number:
MFCD09607735 CAS číslo: 1195995-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...