5-Phenyl-o-phenetidine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Amino-4-ethoxybiphenyl
OR17199-1G
OR17199-250MG
APOSOR17199-1GEA
4630
CZK
APOSOR17199-1G
APOSOR17199-250MG
5-Phenyl-o-phenetidine
5-Phenyl-o-phenetidine
Vzorec:
C₁₄H₁₅NO MW: 213,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425697 CAS číslo: 856343-44-7 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...