Tisk…

1-Boc-3-iodoindole-5-carbonitrile 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Cyano-3-iodo-1H-indole, N-BOC protected

OR1717-1G OR1717-5G
APOSOR1717-1GEA 622 CZK
APOSOR1717-1G APOSOR1717-5G
1-Boc-3-iodoindole-5-carbonitrile 98%
1-Boc-3-iodoindole-5-carbonitrile
Vzorec: C₁₄H₁₃IN₂O₂
MW: 368,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864767
CAS číslo: 864685-26-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR