Tisk…

4,5-Dimethoxy-o-phenylenediamine dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,5-Dimethoxybenzene-1,2-diamine dihydrochloride

OR17128-1G OR17128-250MG
APOSOR17128-1GEA 6020 CZK
APOSOR17128-1G APOSOR17128-250MG
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
Vzorec: C₈H₁₂N₂O₂·2HCl
MW: 241,12 g/mol
Bod tání: 266…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00190674
CAS číslo: 131076-14-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR