Tisk…

4-Amino-5-bromo-2-chloropyridine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine

OR17025-500MG OR17025-2.5G OR17025-10G OR17025-25G
APOSOR17025-500MGEA 2070 CZK
APOSOR17025-500MG APOSOR17025-2.5G APOSOR17025-10G APOSOR17025-25G
4-Amino-5-bromo-2-chloropyridine
4-Amino-5-bromo-2-chloropyridine
Vzorec: C₅H₄BrClN₂
Bod tání: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037459
CAS číslo: 857730-21-3
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR