Tisk…

4-Hydroxy-1H-indole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific
Varování

OR1702-1G OR1702-5G
APOSOR1702-1GEA 3150 CZK
APOSOR1702-1G APOSOR1702-5G
4-Hydroxy-1H-indole-3-carbaldehyde 95%
4-Hydroxy-1H-indole-3-carbaldehyde
Vzorec: C₉H₇NO₂
MW: 161,1574 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864721
CAS číslo: 81779-27-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR