Tisk…

3,5-Dichloro-4-hydroxybenzonitrile 97%

Dodavatel: Apollo Scientific

Synonyms: Chloroxynil

OR17010-1G OR17010-5G
APOSOR17010-1GEA 1780 CZK
APOSOR17010-1G APOSOR17010-5G
3,5-Dichloro-4-hydroxybenzonitrile 97%
3,5-Dichloro-4-hydroxybenzonitrile
Vzorec: C₇H₃Cl₂NO
MW: 188,01 g/mol
Bod tání: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00002177
CAS číslo: 1891-95-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR