Tisk…

2,5-Dichloropyrazine

Dodavatel: Apollo Scientific
Varování

OR16998-1G OR16998-250MG
APOSOR16998-1GEA 1820 CZK
APOSOR16998-1G APOSOR16998-250MG
2,5-Dichloropyrazine
2,5-Dichloropyrazine
Vzorec: C₄H₂Cl₂N₂
MW: 148,9781 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117886
CAS číslo: 19745-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR