Tisk…

4,6-Dihydroxynicotinic acid

Dodavatel: Apollo Scientific

OR16997-1G OR16997-5G
APOSOR16997-1GEA 1910 CZK
APOSOR16997-1G APOSOR16997-5G
4,6-Dihydroxynicotinic acid
4,6-Dihydroxynicotinic acid
Vzorec: C₆H₅NO₄
Bod tání: 309…311 °C
Storage Temperature: Ambient
MDL Number: MFCD00234168
CAS číslo: 5466-62-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR