2-Bromo-3,5-dichloroisonicotinic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-3,5-dichloropyridine-4-carboxylic acid
OR16993-1G
OR16993-250MG
OR16993-5G
APOSOR16993-1GEA
3410
CZK
APOSOR16993-1G
APOSOR16993-250MG
APOSOR16993-5G
2-Bromo-3,5-dichloroisonicotinic acid 96%
2-Bromo-3,5-dichloroisonicotinic acid
Vzorec:
C₆H₂BrCl₂NO₂ MW: 270.90 g/mol Bod tání: 200…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411637 CAS číslo: 343781-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...