α-Bromo-o-toluic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Bromomethyl)benzoic acid
OR16987-1G
OR16987-25G
OR16987-5G
APOSOR16987-1GEA
1660
CZK
APOSOR16987-1G
APOSOR16987-25G
APOSOR16987-5G
α-Bromo-o-toluic acid 97%
α-Bromo-o-toluic acid
Vzorec:
C₈H₇BrO₂ MW: 215,05 g/mol Bod tání: 145…150 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00090420 CAS číslo: 7115-89-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...