Tisk…

Hydroxyterephthalic acid 97%

Dodavatel: Apollo Scientific
Varování

OR16986-1G OR16986-5G OR16986-25G
APOSOR16986-1GEA 1490 CZK
APOSOR16986-1G APOSOR16986-5G APOSOR16986-25G
Hydroxyterephthalic acid 97%
Hydroxyterephthalic acid
Vzorec: C₈H₆O₅
Bod tání: 312…317 °C
Storage Temperature: Ambient
MDL Number: MFCD09835368
CAS číslo: 636-94-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR