Iron(III) citrate hydrate
Dodavatel: Apollo Scientific
|
Varování
|
OR16950-1KG
OR16950-250G
APOSOR16950-1KGEA
4040
CZK
APOSOR16950-1KG
APOSOR16950-250G
Iron(III) citrate hydrate
Iron(III) citrate hydrate
Vzorec:
C₆H₅FeO₇·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149615 CAS číslo: 334024-15-6 EINECS: 222-536-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...