Tisk…

Iodotrimethylsilane

Dodavatel: Apollo Scientific
Nebezpečí

OR16935-100G OR16935-250G
APOSOR16935-100GEA 2480 CZK
APOSOR16935-100G APOSOR16935-250G
Iodotrimethylsilane
Iodotrimethylsilane
Vzorec: C₃H₉ISi
Bod varu: 105…106 °C
Hustota: 1,406 g/cm³ (25 °C)
Bod vzplanutí: –2 °C
Storage Temperature: Freezer
MDL Number: MFCD00001028
CAS číslo: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3+8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR