Tisk…

tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate, 3-(Hydroxymethyl)-5-nitro-1H-indole, N-BOC protected

OR1692-1G OR1692-5G
APOSOR1692-1GEA 870 CZK
APOSOR1692-1G APOSOR1692-5G
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate 98%
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate
Vzorec: C₁₄H₁₆N₂O₅
MW: 292,2872 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864712
CAS číslo: 914349-07-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR