Tisk…

Ethyl [4-(benzyloxy)phenoxy]acetate

Dodavatel: Apollo Scientific

OR16912-10G OR16912-1G OR16912-5G
APOSOR16912-10GEA 14780 CZK
APOSOR16912-10G APOSOR16912-1G APOSOR16912-5G
Ethyl [4-(benzyloxy)phenoxy]acetate
Ethyl [4-(benzyloxy)phenoxy]acetate
Vzorec: C₁₇H₁₈O₄
MW: 286.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03847881
CAS číslo: 142717-44-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR