1-Boc-3-formylindole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1H-Indole-3-carboxaldehyde, N-BOC protected, 1-Boc-3-Formylindole
OR1671-5G
OR1671-10G
OR1671-25G
APOSOR1671-5GEA
622
CZK
APOSOR1671-5G
APOSOR1671-10G
APOSOR1671-25G
1-Boc-3-formylindole
1-Boc-3-formylindole
Vzorec:
C₁₄H₁₅NO₃ Bod tání: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049230 CAS číslo: 57476-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...