Tisk…

1-Oxa-6-azaspiro[3.5]nonane oxalate

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Oxa-6-azaspiro[3.5]nonane hemioxalate

OR16709-1G OR16709-250MG
APOSOR16709-1GEA 9560 CZK
APOSOR16709-1G APOSOR16709-250MG
1-Oxa-6-azaspiro[3.5]nonane oxalate
1-Oxa-6-azaspiro[3.5]nonane oxalate
Vzorec: C₁₆H₂₈N₂O₆
MW: 344,41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27923648
CAS číslo: 1523606-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR