3-Amino-1H-pyrazole-4-carboxamide sulphate(2:1) 97+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Aminopyrazole-4-carboxamide Hemisulphate, 3-Amino-4-pyrazolecarboxamide hemisulphate salt, 3-Amino-4-carbamoylpyrazole Hemisulphate, 3-Amino-1H-pyrazole-4-carboxamide hemisulphate, Allopurinol Related Compound A
OR16674-5G
OR16674-25G
OR16674-100G
APOSOR16674-5GEA
622
CZK
APOSOR16674-5G
APOSOR16674-25G
APOSOR16674-100G
3-Amino-1H-pyrazole-4-carboxamide sulphate(2:1) 97+%
3-Amino-1H-pyrazole-4-carboxamide sulphate(2:1)
Vzorec:
C8H14N8O6S MW: 350,31 g/mol Bod varu: 454,3 °C Bod tání: 224…232 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013093 CAS číslo: 27511-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...