Tisk…

1,11-Undecanedioic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,9-Nonanedicarboxylic acid, 1,9-Nonanedicarboxylicacid, Nonamethylenedicarboxylic acid, Undecanedioic acid

OR16665-100G OR16665-25G
APOSOR16665-100GEA 1450 CZK
APOSOR16665-100G APOSOR16665-25G
1,11-Undecanedioic acid
1,11-Undecanedioic acid
Vzorec: C₁₁H₂₀O₄
MW: 216,28 g/mol
Bod tání: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00004444
CAS číslo: 1852-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR