Tisk…

1,3-Benzenediboronic acid bis(pinacol) ester 97%

Dodavatel: Apollo Scientific
Varování

OR16636-1G OR16636-5G OR16636-25G
APOSOR16636-1GEA 622 CZK
APOSOR16636-1G APOSOR16636-5G APOSOR16636-25G
1,3-Benzenediboronic acid bis(pinacol) ester 97%
1,3-Benzenediboronic acid bis(pinacol) ester
Vzorec: C₁₈H₂₈B₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09864185
CAS číslo: 196212-27-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR