Tisk…

1,4-Benzodioxane-6-boronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 1,4-Benzodioxane-6-boronic acid, pinacol ester, 2,3-Dihydro-1,4-benzodioxine-6-boronic acid, pinacol ester, 2-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR16635-1G OR16635-5G
APOSOR16635-1GEA 2400 CZK
APOSOR16635-1G APOSOR16635-5G
1,4-Benzodioxane-6-boronic acid pinacol ester
1,4-Benzodioxane-6-boronic acid pinacol ester
Vzorec: C₁₄H₁₉BO₄
MW: 262,1093 g/mol
Bod tání: 54…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663890
CAS číslo: 517874-21-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR