Tisk…

1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,3-Dimethylpyrazole-4-boronic acid Pinacol ester, 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 2-(1,3-Dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR16623-1G OR16623-500MG OR16623-5G
APOSOR16623-1GEA 2070 CZK
APOSOR16623-1G APOSOR16623-500MG APOSOR16623-5G
1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Vzorec: C₁₁H₁₉BN₂O₂
MW: 222,0918 g/mol
Bod tání: 70…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09864190
CAS číslo: 1046832-21-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR