tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Cyano-1H-indole-3-carboxaldehyde, N-BOC protected
OR1657-1G
OR1657-5G
OR1657-25G
APOSOR1657-1GEA
4200
CZK
APOSOR1657-1G
APOSOR1657-5G
APOSOR1657-25G
tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-5-cyano-3-formyl-1H-indole-1-carboxylate
Vzorec:
C₁₅H₁₄N₂O₃ Storage Temperature: Refrigerator |
CAS číslo:
914348-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...