Tisk…

1-Boc-4-bromoindole 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Boc-4-Bromoindole, 4-Bromo-1H-indole, N-BOC protected

OR1655-1G OR1655-500MG
APOSOR1655-1GEA 1040 CZK
APOSOR1655-1G APOSOR1655-500MG
1-Boc-4-bromoindole 95%
1-Boc-4-bromoindole
Vzorec: C₁₃H₁₄BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD04117874
CAS číslo: 676448-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR