(±)-Tetrahydrofuran-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tetrahydrofuran-3-carboxylic acid, Tetrahydro-3-furoic acid
OR16452-1G
OR16452-5G
OR16452-25G
APOSOR16452-1GEA
622
CZK
APOSOR16452-1G
APOSOR16452-5G
APOSOR16452-25G
(±)-Tetrahydrofuran-3-carboxylic acid
(±)-Tetrahydrofuran-3-carboxylic acid
Vzorec:
C₅H₈O₃ MW: 116,1152 g/mol Bod varu: 140…141 °C/15mmHg Hustota: 1,214 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075173 CAS číslo: 89364-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...